Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Multichannel Pump Head for Precision Tubing; 8-Channel
Catalog No. NC0683147
Encompass
The L/S multichannel pump heads are designed with machined stainless steel (SS) and anodized aluminum with SS rotor, bearings, and rollers for reliable, continuous-duty operation. Choose pump heads for microbore tubing, L/S Precision tubing, or L/S High-Performance Precision tubing based on your application requirements. Compatible with L/S drives that accept two or more pump heads and have a 1/10 hp minimum.
Catalog No. NC0683147 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0753508
The L/S multichannel pump heads are designed with machined stainless steel (SS) and anodized aluminum with SS rotor, bearings, and rollers for reliable, continuous-duty operation. Choose pump heads for microbore tubing, L/S Precision tubing, or L/S High-Performance Precision tubing based on your application requirements. Compatible with L/S drives that accept two or more pump heads and have a 1/10 hp minimum.