Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Miniflex OEM Pump Head, Single-Channel
Catalog No. NC0996925
Encompass
The Masterflex Miniflex pump heads feature a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life. A compact overall size envelope makes these heads ideal for OEM applications and integration into fluid systems and machinery.
Catalog No. NC0996925 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7722000
The Masterflex Miniflex pump heads feature a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life. A compact overall size envelope makes these heads ideal for OEM applications and integration into fluid systems and machinery.