Learn More
Accuflow Systems Precision Digital Drive Motor With Graphical Interface, Masterflex, L/S, Easy-Load, Ii Pump Head (Thin Wall Tubing), Flow Rates Of 0.001-3400 Ml/Min
Catalog No. NC1475875
Encompass_Preferred
This precision digital drive and pump head offers a maintenance-free high-accuracy motor and a graphical interface – makes setup and operation easier than ever! Flow rates of 0.001 to 3400 mL/min with L/S tubing. Flow rate depends on drive rpm and tubing size. Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually. This pump drive includes a 6-ft (1.8m) line cord with IEC 320/CEE22 socket and are shipped with country specific plug/cord set as well as a Masterflex™ L/S™ Easy-Load™ II Pump Head featuring automatic retention which holds precision (thin-walled) tubing securely in place without manual adjustment. Just load the tubing and start the pump.
Catalog No. NC1475875 Supplier Accuflow Systems Supplier No. 7771251
Add to Cart
Add to Cart
Request a Quote
This precision digital drive and pump head offers a maintenance-free high-accuracy motor and a graphical interface – makes setup and operation easier than ever! Flow rates of 0.001 to 3400 mL/min with L/S tubing. Flow rate depends on drive rpm and tubing size. Graphical LCD shows four operating modes continuous run, timed dispense, copy dispense, volume dispense. View pump performance continually. This pump drive includes a 6-ft (1.8m) line cord with IEC 320/CEE22 socket and are shipped with country specific plug/cord set as well as a Masterflex™ L/S™ Easy-Load™ II Pump Head featuring automatic retention which holds precision (thin-walled) tubing securely in place without manual adjustment. Just load the tubing and start the pump.