Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Variable-Speed Air-Powered Drive, 60 to 600 rpm
Catalog No. NC9006723
Encompass
Use this variable-speed air-powered drive whenever electrical power is unsafe or impractical. Careful grounding to protect from static electricity is vital for safe operation. Drive accepts nine pump head types: Standard, Easy-Load, Easy-Load II, Easy-Load 3, Multichannel, High-Performance, Cartridge, PTFE-Tubing, and PTFE-Diaphragm.
Catalog No. NC9006723 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0756900
Use this variable-speed air-powered drive whenever electrical power is unsafe or impractical. Careful grounding to protect from static electricity is vital for safe operation. Drive accepts nine pump head types: Standard, Easy-Load, Easy-Load II, Easy-Load 3, Multichannel, High-Performance, Cartridge, PTFE-Tubing, and PTFE-Diaphragm.