Learn More
AVANTOR MASTERFLEX (VWR LLC) Variable-Speed Pump System, 600 rpm, 115 VAC
Catalog No. NC1561113
Encompass
Masterflex 77910-50 L/S Variable-Speed Pump System with Easy-Load II Thick-Wall Pump Head, 600 rpm, 115 VAC, Compact, cost effective pump systems for general fluid transfer applicationsSimple operation with single-turn speed controlSeparate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power onReversible motor lets you easily prime/purge tubing and pump in either directionRemote control of start/stop via DB9 connector on back of drive
Catalog No. NC1561113 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7791050
Masterflex 77910-50 L/S Variable-Speed Pump System with Easy-Load II Thick-Wall Pump Head, 600 rpm, 115 VAC, Compact, cost effective pump systems for general fluid transfer applicationsSimple operation with single-turn speed controlSeparate power switch enables you to maintain speed setting when turning drive on/off; front-panel LED indicates power onReversible motor lets you easily prime/purge tubing and pump in either directionRemote control of start/stop via DB9 connector on back of drive