Learn More
Medchemexpress LLC L-selectin/CD62L Cy 10ug | 10ug
SDP
Catalog No. 5000186649
Encompass_Preferred
L-selectin/CD62L is a calcium-dependent lectin that aids cell adhesion by binding to neighboring glycoproteins It mediates lymphocyte adhesion to endothelial cells in peripheral lymph nodes causing leukocytes to tether and roll L-selectin/CD62L Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived L-selectin/CD62L protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86649 Supplier Medchemexpress LLC Supplier No. HYP7748310UG
L-selectin/CD62L is a calcium-dependent lectin that aids cell adhesion by binding to neighboring glycoproteins It mediates lymphocyte adhesion to endothelial cells in peripheral lymph nodes causing leukocytes to tether and roll L-selectin/CD62L Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived L-selectin/CD62L protein expressed by HEK293 with C-His labeled tag