Learn More
Medchemexpress LLC L-selectin/CD62L Cy 50ug | 50ug
SDP
Catalog No. 5000187104
Encompass_Preferred
L-selectin/CD62L is a calcium-dependent lectin that aids cell adhesion by binding to neighboring glycoproteins It mediates lymphocyte adhesion to endothelial cells in peripheral lymph nodes causing leukocytes to tether and roll L-selectin/CD62L Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived L-selectin/CD62L protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87104 Supplier Medchemexpress LLC Supplier No. HYP7748350UG
L-selectin/CD62L is a calcium-dependent lectin that aids cell adhesion by binding to neighboring glycoproteins It mediates lymphocyte adhesion to endothelial cells in peripheral lymph nodes causing leukocytes to tether and roll L-selectin/CD62L Protein Cynomolgus (HEK293 His) is the recombinant cynomolgus-derived L-selectin/CD62L protein expressed by HEK293 with C-His labeled tag