Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-selectin/CD62L protein, mouse (HEK293, His) | ≥98.0% | 100 UG
SDP

Catalog No. 5000294536
Encompass_Preferred

Recombinant mouse L-selectin (CD62L) ectodomain expressed in HEK293 cells and produced with a C-terminal His tag. Supplied as a sterile, 0.22 μm-filtered PBS solution for in vitro research use, it is suitable for binding and receptor-ligand interaction studies, antibody development, and biochemical assays. Consult the product COA and datasheet for batch-specific concentration and analytical data.

  • Expressed in HEK293 cells.
  • C-terminal His tag for detection or purification.
  • Molecular weight approximately 70-80 kDa.
  • Purity ≥98.0% (per COA).
  • Endotoxin <1 EU/μg.
  • Formulated in PBS, 0.22 μm filtered.
  • Store at -80°C; ship on dry ice; avoid repeated freeze-thaw cycles.

Catalog No. 50-002-94536 Supplier Medchemexpress LLC Supplier No. HYP74766100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse L-selectin (CD62L) ectodomain expressed in HEK293 cells and produced with a C-terminal His tag. Supplied as a sterile, 0.22 μm-filtered PBS solution for in vitro research use, it is suitable for binding and receptor-ligand interaction studies, antibody development, and biochemical assays. Consult the product COA and datasheet for batch-specific concentration and analytical data.

  • Expressed in HEK293 cells.
  • C-terminal His tag for detection or purification.
  • Molecular weight approximately 70-80 kDa.
  • Purity ≥98.0% (per COA).
  • Endotoxin <1 EU/μg.
  • Formulated in PBS, 0.22 μm filtered.
  • Store at -80°C; ship on dry ice; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.