Learn More
Medchemexpress LLC L-selectin/CD62L protein, mouse (HEK293, His) | ≥98.0% | 50 UG

Supplier: Medchemexpress LLC HYP7476650UG
Recombinant mouse L-selectin (CD62L) is the extracellular domain of the protein expressed in HEK293 cells with a C-terminal His tag. It is supplied as a 0.22 μm filtered PBS solution (pH 7.4) at 0.34 mg/mL, with an approximate molecular weight of 70-80 kDa and high purity suitable for research applications in immunology and cell adhesion studies.
- Recombinant mouse extracellular domain suitable for in vitro assays.
- Expressed in HEK293 for mammalian-like post-translational modifications.
- C-terminal His tag facilitates purification and detection.
- Supplied as a filtered PBS solution ready for dilution or use.
- High purity (≥98.0%) supports biochemical and binding studies.
- Concentration provided for convenient aliquoting and dosing.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.