Learn More
Medchemexpress LLC L-Selenocystine | 29621-88-3 | 98.0% | 250 MG
SDP
Catalog No. 5000341483
Encompass_Preferred
L-Selenocystine is a selenium-containing amino acid known for its redox properties. It demonstrates cytotoxic effects on various tumor cells, promoting the production of reactive oxygen species (ROS) and apoptosis. This compound also has the ability to inhibit the Nrf2 and autophagy pathways, contributing to its anti-tumor activity.
  • Selenium-containing amino acid
  • Exhibits redox properties
  • Cytotoxic to various tumor cells
  • Induces ROS production and apoptosis
  • Blocks Nrf2 and autophagy pathways
  • Demonstrates anti-tumor activity
  • Inhibits proliferation of HepG2 cells
  • Modulates apoptosis-related proteins
Catalog No. 50-003-41483 Supplier Medchemexpress LLC Supplier No. 250MGHY129960
L-Selenocystine is a selenium-containing amino acid known for its redox properties. It demonstrates cytotoxic effects on various tumor cells, promoting the production of reactive oxygen species (ROS) and apoptosis. This compound also has the ability to inhibit the Nrf2 and autophagy pathways, contributing to its anti-tumor activity.
  • Selenium-containing amino acid
  • Exhibits redox properties
  • Cytotoxic to various tumor cells
  • Induces ROS production and apoptosis
  • Blocks Nrf2 and autophagy pathways
  • Demonstrates anti-tumor activity
  • Inhibits proliferation of HepG2 cells
  • Modulates apoptosis-related proteins