Learn More
Medchemexpress LLC L-SelenoMethionine | 3211-76-5 | 99.8% | 10 MM 1 ML
SDP
Catalog No. 5000407502
Encompass_Preferred
L-SelenoMethionine, an L-isomer of Selenomethionine, is a major natural food-form of selenium. It is a cancer chemopreventive agent that can reduce cancer incidence through dietary supplementation and induce apoptosis of cancer cells. It can also increase the expression of glutathione peroxidase.
  • Selectively inhibits the growth of prostate cancer cells compared to normal cells (1-500 μM; 24-72 h).
  • Induces apoptosis in prostate cancer cells (500 μM; 48 h).
  • Causes an increase in arrest in the G2-M phase of the cell cycle selectively in prostate cancer cells (500 μM; 48 h).
  • Mitigates gene expression associated with the cellular stress response from 10 cGy irradiation (5 μM; 24 h).
  • Partially or completely prevents the decrease in serum or plasma levels of total antioxidants in rats exposed to gamma rays, protons, or HZE particles (0.06-12 μg/g diet; p.o. for 3 d).
Catalog No. 50-004-07502 Supplier Medchemexpress LLC Supplier No. HYB1000A10MM/1ML
L-SelenoMethionine, an L-isomer of Selenomethionine, is a major natural food-form of selenium. It is a cancer chemopreventive agent that can reduce cancer incidence through dietary supplementation and induce apoptosis of cancer cells. It can also increase the expression of glutathione peroxidase.
  • Selectively inhibits the growth of prostate cancer cells compared to normal cells (1-500 μM; 24-72 h).
  • Induces apoptosis in prostate cancer cells (500 μM; 48 h).
  • Causes an increase in arrest in the G2-M phase of the cell cycle selectively in prostate cancer cells (500 μM; 48 h).
  • Mitigates gene expression associated with the cellular stress response from 10 cGy irradiation (5 μM; 24 h).
  • Partially or completely prevents the decrease in serum or plasma levels of total antioxidants in rats exposed to gamma rays, protons, or HZE particles (0.06-12 μg/g diet; p.o. for 3 d).