Learn More
TARGETMOL CHEMICALS INC L-SelenoMethionine 500MG
SDP
Catalog No. 502879665
Encompass_Preferred
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. L-SelenoMethionine (L-(+)-Selenomethionine) is a naturally occurring amino acid. It promotes cell cycle progression and elevates the expression of the antioxidant enzymes thioredoxin reductase, glutathione reductase. Purity 98.55%
Catalog No. 50-287-9665 Supplier TARGETMOL CHEMICALS INC Supplier No. T5372500MG
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. L-SelenoMethionine (L-(+)-Selenomethionine) is a naturally occurring amino acid. It promotes cell cycle progression and elevates the expression of the antioxidant enzymes thioredoxin reductase, glutathione reductase. Purity 98.55%