Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Serinamide hydrochloride, 98%, Thermo Scientific™

Catalog No. AC303162500
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303162500 250 mg Glass Bottle
AC303160010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC303162500 Supplier Thermo Scientific Chemicals Supplier No. 303162500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65414-74-6
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00067560
InChI Key VURWDDZIWBGXCK-DKWTVANSSA-N
Synonym l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl
PubChem CID 16212403
IUPAC Name (2S)-2-amino-3-hydroxypropanamide;hydrochloride
SMILES C(C(C(=O)N)N)O.Cl

Specifications

Melting Point 184°C to 190°C
Color Purple to Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOCHCH(NH)CONH·HCl
Specific Rotation +13.5° to +15.5° (20°C, 589nm) (c=1, H2O)
Specific Rotation Condition +14.5° (20°C c=1,H2O)
Quantity 250 mg
Formula Weight 140.57
Percent Purity ≥97.5% (Argentometry)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Serinamide hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.