Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Serine-1,2,3-13C3 | 201595-68-8 | MFCD00144677 | 108.07 | 25 MG
SDP

Catalog No. 5000389317
Encompass_Preferred

L-Serine-13C3 is the 13C-labeled L-Serine, a non-essential amino acid that plays a central role in cellular proliferation. It is intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.77%.
  • Appearance: Solid.
  • Color: White to off-white.

Catalog No. 50-003-89317 Supplier Medchemexpress LLC Supplier No. 25MGHYN0650S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Serine-13C3 is the 13C-labeled L-Serine, a non-essential amino acid that plays a central role in cellular proliferation. It is intended for research use only.

  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Purity: 99.77%.
  • Appearance: Solid.
  • Color: White to off-white.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.