Learn More
Medchemexpress LLC L-Serine-13C | 89232-77-9 | 106.09 | 1 MG
SDP
Catalog No. 5000388678
Encompass_Preferred
L-Serine-13C is the 13C-labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88678 Supplier Medchemexpress LLC Supplier No. HYN0650S11MG
L-Serine-13C is the 13C-labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.