Learn More
Medchemexpress LLC L-Serine-13C3,15N | 202407-34-9 | 109.06 | 5 MG
SDP
Catalog No. 5000388531
Encompass_Preferred
L-Serine-13C3,15N is the 13C- and 15N-labeled L-Serine, a non-essential amino acid playing a central role in cellular proliferation. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88531 Supplier Medchemexpress LLC Supplier No. HYN0650S55MG
L-Serine-13C3,15N is the 13C- and 15N-labeled L-Serine, a non-essential amino acid playing a central role in cellular proliferation. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.