Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-serine-13C3,15N | 202407-34-9 | MFCD00144678 | 99.9% | 109.06 | 13C3H7 15NO3 | 10 MG
SDP

Catalog No. 5000296839
Encompass_Preferred

L-Serine-13C3,15N is a stable isotope-labeled L-serine in solid form, with three 13C atoms and one 15N atom. It is supplied as a high-purity analytical standard for mass spectrometry, metabolic tracing, and biochemical assays.

  • Stable isotope-labeled reference standard for MS and tracer studies.
  • Isotopic enrichment: three 13C atoms and one 15N atom.
  • High purity (99.9%) suitable for quantitative analysis.
  • Molecular weight 109.06 and CAS number 202407-34-9 for unambiguous identification.
  • Physical form: solid powder; soluble in water.
  • Typical applications include internal standard, metabolic flux analysis, and pathway tracing.

Catalog No. 50-002-96839 Supplier Medchemexpress LLC Supplier No. HYN0650S510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Serine-13C3,15N is a stable isotope-labeled L-serine in solid form, with three 13C atoms and one 15N atom. It is supplied as a high-purity analytical standard for mass spectrometry, metabolic tracing, and biochemical assays.

  • Stable isotope-labeled reference standard for MS and tracer studies.
  • Isotopic enrichment: three 13C atoms and one 15N atom.
  • High purity (99.9%) suitable for quantitative analysis.
  • Molecular weight 109.06 and CAS number 202407-34-9 for unambiguous identification.
  • Physical form: solid powder; soluble in water.
  • Typical applications include internal standard, metabolic flux analysis, and pathway tracing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.