Learn More
Medchemexpress LLC L-Serine-13C3,15N,d3 | 1994299-33-0 | 112.08 | 5 MG
SDP
Catalog No. 5000389118
Encompass_Preferred
L-Serine-13C3, 15N, d3 is the deuterium, 13C-, and 15-labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium, 13C-, and 15-labeled L-Serine
  • Plays a central role in cellular proliferation
  • Used as tracers for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-89118 Supplier Medchemexpress LLC Supplier No. HYN0650S35MG
L-Serine-13C3, 15N, d3 is the deuterium, 13C-, and 15-labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium, 13C-, and 15-labeled L-Serine
  • Plays a central role in cellular proliferation
  • Used as tracers for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs