Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-serine (13C3,15N,d3) | 1994299-33-0 | 99.6% | 112.08 g/mol | ¹³C₃H₄D₃¹⁵NO₃ | 10 MG
SDP

Catalog No. 5000297275
Encompass_Preferred

Isotopically labeled L-serine for use as a tracer or internal standard in mass spectrometry and metabolic studies. Supplied for research use, the compound contains 13C, 15N, and deuterium labels and is characterized by high chemical purity.

  • Isotopically labeled amino acid (13C3, 15N, d3)
  • Intended for tracer studies and internal standard use in MS
  • Purity 99.6% (GC)
  • Molecular weight 112.08 g/mol
  • CAS number 1994299-33-0
  • Provided as a small research quantity suitable for analytical use

Catalog No. 50-002-97275 Supplier Medchemexpress LLC Supplier No. HYN0650S310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isotopically labeled L-serine for use as a tracer or internal standard in mass spectrometry and metabolic studies. Supplied for research use, the compound contains 13C, 15N, and deuterium labels and is characterized by high chemical purity.

  • Isotopically labeled amino acid (13C3, 15N, d3)
  • Intended for tracer studies and internal standard use in MS
  • Purity 99.6% (GC)
  • Molecular weight 112.08 g/mol
  • CAS number 1994299-33-0
  • Provided as a small research quantity suitable for analytical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.