Learn More
Medchemexpress LLC L-Serine-13C3 | 201595-68-8 | 108.07 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0650S1MG
Encompass_Preferred
L-Serine-13C3 is the 13C-labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89767