Learn More
Medchemexpress LLC L-Serine-13C3 | 201595-68-8 | 108.07 | 100 MG
SDP
Catalog No. 5000388679
Encompass_Preferred
L-Serine-13C3 is a 13C-labeled version of L-Serine, a non-essential amino acid essential for cellular proliferation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88679 Supplier Medchemexpress LLC Supplier No. HYN0650S100MG
L-Serine-13C3 is a 13C-labeled version of L-Serine, a non-essential amino acid essential for cellular proliferation.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.