Learn More
Medchemexpress LLC L-Serine-15N | 59935-32-9 | 106.09 | 50 MG
SDP
Catalog No. 5000388645
Encompass_Preferred
L-Serine-15N is the 15N-labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-88645 Supplier Medchemexpress LLC Supplier No. HYN0650S1050MG
L-Serine-15N is the 15N-labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.