Learn More
AJINOMOTO USA INC L-Serine (L-Ser) 1Kg
Catalog No. NC1426899
Encompass
Chemical Name: (S)-2-Amino-3-hydroxypropionic Acid; Chemical Formula: C3H7NO3; Molecular Weight: 105.0900; Nitrogen Content: 0.13330000 CAS Number: 56-45-1; Assay: 98.5% to 101.0%; Description: White crystals or crystalline powder; slightly sweet taste 1Kg
Catalog No. NC1426899 Supplier AJINOMOTO USA INC Supplier No. 34251
Chemical Name: (S)-2-Amino-3-hydroxypropionic Acid; Chemical Formula: C3H7NO3; Molecular Weight: 105.0900; Nitrogen Content: 0.13330000 CAS Number: 56-45-1; Assay: 98.5% to 101.0%; Description: White crystals or crystalline powder; slightly sweet taste 1Kg