Learn More
Medchemexpress LLC L-Serine-2,3,3-d3 | 105591-10-4 | 108.11 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0650S81MG
Encompass_Preferred
L-Serine-d3 is the deuterium labeled L-Serine, a non-essential amino acid that plays a central role in cellular proliferation. It is suitable for use as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis
  • Supports cellular proliferation studies
Catalog No. 50-003-89063