Learn More
AJINOMOTO USA INC L-SERINE 50KG
Supplier:  AJINOMOTO USA INC 21018
Encompass
Chemical Name: (S)-2-Amino-3-hydroxypropionic AcidChemical Formula: C3H7NO3 Molecular Weight: 105.0900 Nitrogen Content: 0.13330000 CAS Number: 56-45-1Assay: 98.5% to 101.0% Description: White crystals or crystalline powder; slightly sweet tast
Catalog No. NC2591697