Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Serine, 98.5%, Spectrum™ Chemical
SDP

Catalog No. 18611387 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-387 25 g Poly Pail
18-611-386 100 g Poly Pail
18-611-385 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-387 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SE12525GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56-45-1 | 105.09 g/mol

Chemical Identifiers

CAS 56-45-1

Specifications

CAS Max % 1
Lead (Pb) 10 ppm
Loss on Drying 0.003
Packaging Poly Pail
Specific Rotation +13.6° to +15.6°
Molecular Weight (g/mol) 105.09
Quantity 25 g
Percent Purity 98.5%
Grade Reagent
Chemical Name or Material L-Serine

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.