Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Serine, ≥99%, MP Biomedicals™

Catalog No. ICN10287305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10287305 5 g
ICN10287380 100 g
ICN10287325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10287305 Supplier MP Biomedicals, Inc Supplier No. 0210287305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064224
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 5 g
Formula Weight 105.09
Physical Form Free-flowing Powder
Chemical Name or Material L-Serine
EINECSNumber 200-274-3
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.