Learn More
Medchemexpress LLC L-Serine-d2 | 95034-57-4 | 107.10 | 100 MG
SDP
Catalog No. 5000388955
Encompass_Preferred
L-Serine-d2 is the deuterium labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-88955 Supplier Medchemexpress LLC Supplier No. HYN0650S7100MG
L-Serine-d2 is the deuterium labeled L-Serine. L-Serine ((-)-Serine; (S)-Serine), one of the so-called non-essential amino acids, plays a central role in cellular proliferation.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS