Learn More
Medchemexpress LLC L-Serine-d3 | 105591-10-4 | 108.11 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYN0650S810MM/1ML
Encompass_Preferred
L-Serine-d3 is the deuterium-labeled L-Serine, a non-essential amino acid crucial for cellular proliferation. This product can be used as a tracer or as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS techniques.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS applications
  • Plays a central role in cellular proliferation
Catalog No. 50-003-88481