Learn More
Medchemexpress LLC L-serine-d7 | 935275-35-7 | 100.0% | 112.14 g·mol⁻1 | C3D7NO3 | 10 MG
SDP
Catalog No. 5000298067
Encompass_Preferred
L-Serine-d7 is the deuterium (d7) isotopically labeled form of L-serine, provided as a high-purity research-grade standard for analytical and biochemical applications. Molecular weight 112.14 g·mol⁻1; CAS 935275-35-7.
  • High isotopic enrichment (d7) for accurate mass spectrometry quantitation.
  • High purity: 99.97%.
  • Suitable as an internal standard for LC-MS/MS.
  • Useful for metabolic tracing and isotopic labeling studies.
  • Supplied as a 10 mg solid, convenient for small-scale analyses.
Catalog No. 50-002-98067 Supplier Medchemexpress LLC Supplier No. HYN0650S1110MG
L-Serine-d7 is the deuterium (d7) isotopically labeled form of L-serine, provided as a high-purity research-grade standard for analytical and biochemical applications. Molecular weight 112.14 g·mol⁻1; CAS 935275-35-7.
  • High isotopic enrichment (d7) for accurate mass spectrometry quantitation.
  • High purity: 99.97%.
  • Suitable as an internal standard for LC-MS/MS.
  • Useful for metabolic tracing and isotopic labeling studies.
  • Supplied as a 10 mg solid, convenient for small-scale analyses.