Learn More
Medchemexpress LLC L-serine, ethyl ester hydrochloride | 26348-61-8 | MFCD00012594 | >98.0% | 169.61 | C5H12ClNO3 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW016996500G
Encompass_Preferred
H-Ser-OEt·HCl (L-serine, ethyl ester hydrochloride) is an amino acid derivative supplied as a white to off-white solid for research use. It is used as a synthetic building block in peptide chemistry and biochemical studies, offering an ethyl ester hydrochloride form of serine that aids handling and reaction compatibility.
  • High purity suitable for analytical and synthetic applications.
  • White to off-white solid physical form.
  • Provides an ethyl ester hydrochloride derivative of serine for peptide synthesis.
  • Stable when stored sealed at 4°C and protected from moisture.
  • Soluble in common aqueous and organic solvents for reaction use.
Catalog No. 50-003-00434