Learn More
Medchemexpress LLC L-Serine ethyl ester hydrochloride | 26348-61-8 | MFCD00012594 | ≥98.0% | 169.61 g/mol | C5H12ClNO3 | 100 G
SDP
Catalog No. 5000301060
Encompass_Preferred
L-Serine ethyl ester hydrochloride is the ethyl ester hydrochloride salt of the amino acid L-serine. Supplied as a white to off-white solid with high purity, it is used as an amino acid derivative in peptide synthesis, biochemical studies, and related research applications.
  • High purity ≥98.0% (NMR).
  • White to off-white solid for easy handling and weighing.
  • Suitable for peptide synthesis and biochemical studies.
  • Hydrochloride salt form to improve solubility in polar solvents.
  • Recommended storage at 4°C, protected from moisture.
Catalog No. 50-003-01060 Supplier Medchemexpress LLC Supplier No. HYW016996100G
L-Serine ethyl ester hydrochloride is the ethyl ester hydrochloride salt of the amino acid L-serine. Supplied as a white to off-white solid with high purity, it is used as an amino acid derivative in peptide synthesis, biochemical studies, and related research applications.
  • High purity ≥98.0% (NMR).
  • White to off-white solid for easy handling and weighing.
  • Suitable for peptide synthesis and biochemical studies.
  • Hydrochloride salt form to improve solubility in polar solvents.
  • Recommended storage at 4°C, protected from moisture.