Learn More
Medchemexpress LLC L-Serine hydrochloride | 16428-75-4 | MFCD03791087 | 97.0% | 141.55 g/mol | C3H8ClNO3 | 250 MG
SDP
Catalog No. 5000283709
Encompass_Preferred
L-Serine hydrochloride is the hydrochloride salt (1:1) of the amino acid L-serine supplied as a white to off-white crystalline powder for research use. It is commonly used as a biochemical reagent, media supplement, and synthetic building block in enzymology and cell biology studies. The material has a molecular weight of 141.55 g/mol and CAS 16428-75-4, with typical purity around 97% and recommended storage dry at room temperature.
  • High-purity reagent suitable for biochemical research.
  • Hydrochloride form improves aqueous solubility.
  • Useful as a media supplement and enzyme assay reagent.
  • White to off-white crystalline powder, easy to weigh and dissolve.
  • Stable when stored dry at room temperature.
Catalog No. 50-002-83709 Supplier Medchemexpress LLC Supplier No. HYW142089250MG
L-Serine hydrochloride is the hydrochloride salt (1:1) of the amino acid L-serine supplied as a white to off-white crystalline powder for research use. It is commonly used as a biochemical reagent, media supplement, and synthetic building block in enzymology and cell biology studies. The material has a molecular weight of 141.55 g/mol and CAS 16428-75-4, with typical purity around 97% and recommended storage dry at room temperature.
  • High-purity reagent suitable for biochemical research.
  • Hydrochloride form improves aqueous solubility.
  • Useful as a media supplement and enzyme assay reagent.
  • White to off-white crystalline powder, easy to weigh and dissolve.
  • Stable when stored dry at room temperature.