Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Serine, MP Biomedicals™

Catalog No. MP210287391 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210287391 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210287391 Supplier MP Biomedicals, Inc Supplier No. 0210287391
Add to Cart
Add to Cart

Functions as a precursor of glycine by serine hydroxymethyl transferase. L-Serine, MP Biomedicals is one of the two biological amino acids with a hydroxyl substituted side chain. It is hydrophilic in character.

L-serine is used in the study of influence of pulsed applications of serine on polyglucose synthesis by Fusobacterium nucleatum cultures. Supplementation of minimal media with serine has been shown to restore growth in Escherichia coli that lack the PII and GlnK genes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 222°C (decomposition)(literature)
Quantity 1 kg
Solubility Information Soluble in water (50mg/mL); insoluble in alcohol and ether.
Optical Rotation 10–20° (c=10, 2N HCl)
Formula Weight 105.09
Percent Purity ≥99%
Physical Form Free-flowing Powder
Recommended Storage Room Temperature (15–30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.