Learn More
Medchemexpress LLC L-Serine, N-[[5-(dimethylamino)-1-naphthalenyl]sulfonyl]glycyl-L-cysteinyl-L-valyl-L-leucyl- (TFA) | 143744-88-1 | 99.7% | 710.86 | 5 MG
SDP
Catalog No. 5000399597
Encompass_Preferred
Dansyl-Gly-Cys-Val-Leu-Ser TFA is a dansyl-labeled peptide that functions as a fluorescent dye. This product appears as a white to off-white solid, and its mass spectrometry data is consistent with its structure, ensuring compliance with specifications.
  • Functions as a fluorescent dye
  • Dansyl-labeled peptide
  • White to off-white solid appearance
  • MS consistent with structure
  • Complies with specifications
Catalog No. 50-003-99597 Supplier Medchemexpress LLC Supplier No. HYP4337A5MG
Dansyl-Gly-Cys-Val-Leu-Ser TFA is a dansyl-labeled peptide that functions as a fluorescent dye. This product appears as a white to off-white solid, and its mass spectrometry data is consistent with its structure, ensuring compliance with specifications.
  • Functions as a fluorescent dye
  • Dansyl-labeled peptide
  • White to off-white solid appearance
  • MS consistent with structure
  • Complies with specifications