Learn More
Medchemexpress LLC L-Serine-N,N,O,1,2,3,3-d7 | 935275-35-7 | 112.14 | 50 MG
SDP
Catalog No. 5000388523
Encompass_Preferred
L-Serine-d7 is the deuterium-labeled form of L-Serine, a non-essential amino acid crucial for cellular proliferation. This compound is ideal for use in various research applications, particularly as a tracer or internal standard.
  • Can be utilized as a tracer in research applications.
  • Serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Deuterium substitution may influence the pharmacokinetic and metabolic profiles of drugs.
  • Boasts a purity of 99.97%.
  • Presented as a white to off-white solid.
  • Soluble in H2O at 50 mg/mL (445.87 mM), requiring ultrasonic treatment for dissolution.
Catalog No. 50-003-88523 Supplier Medchemexpress LLC Supplier No. HYN0650S1150MG
L-Serine-d7 is the deuterium-labeled form of L-Serine, a non-essential amino acid crucial for cellular proliferation. This compound is ideal for use in various research applications, particularly as a tracer or internal standard.
  • Can be utilized as a tracer in research applications.
  • Serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Deuterium substitution may influence the pharmacokinetic and metabolic profiles of drugs.
  • Boasts a purity of 99.97%.
  • Presented as a white to off-white solid.
  • Soluble in H2O at 50 mg/mL (445.87 mM), requiring ultrasonic treatment for dissolution.