Learn More
Medchemexpress LLC L-Seryl-L-isoleucyl-L-isoleucyl-L-asparaginyl-L-phenylalanyl-L-a-glutamyl-L-lysyl-L-leucine trifluoroacetate | 1262751-08-5 | 99.9% | 1077.15 | 25 MG
SDP
Catalog No. 5000426561
Encompass_Preferred
OVA Peptide(257-264) is a class I (Kb)-restricted peptide epitope of OVA. This octameric peptide, derived from ovalbumin, can be presented by the class I MHC molecule, H-2Kb.
  • Class I (Kb)-restricted peptide epitope of OVA.
  • Octameric peptide derived from ovalbumin.
  • Presented by the class I MHC molecule, H-2Kb.
  • Protein context influences TAP-independent processing for MHC class I presentation.
  • Presented with differential dependence on the TAP transporter.
  • Contained within MBPCrl-OVA or Crl-OVA bacterial fusion proteins is presented with little dependence on the TAP transporter.
  • Contained within full-length ovalbumin is largely dependent on the TAP transporter.
Catalog No. 50-004-26561 Supplier Medchemexpress LLC Supplier No. 25MGHYP1489A
OVA Peptide(257-264) is a class I (Kb)-restricted peptide epitope of OVA. This octameric peptide, derived from ovalbumin, can be presented by the class I MHC molecule, H-2Kb.
  • Class I (Kb)-restricted peptide epitope of OVA.
  • Octameric peptide derived from ovalbumin.
  • Presented by the class I MHC molecule, H-2Kb.
  • Protein context influences TAP-independent processing for MHC class I presentation.
  • Presented with differential dependence on the TAP transporter.
  • Contained within MBPCrl-OVA or Crl-OVA bacterial fusion proteins is presented with little dependence on the TAP transporter.
  • Contained within full-length ovalbumin is largely dependent on the TAP transporter.