Learn More
Bellco Glass, Inc. L-Shape Sparge W/Stainless Steel Frit,15L For 10mm Port
SDP
Catalog No. 501946478
Encompass_Preferred
Stainless stealL-Shaped Sparage With SST Frit has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 10mm port on 15L metric headplate.. MADE IN THE USA
Catalog No. 50-194-6478 Supplier Bellco Glass, Inc. Supplier No. 790915052
Stainless stealL-Shaped Sparage With SST Frit has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 10mm port on 15L metric headplate.. MADE IN THE USA