Learn More
Bellco Glass, Inc. L-Shape Sparger W/Stainless Steel Frit ,3L For 10mm Port
SDP
Catalog No. 501946435
Encompass_Preferred
Stainless steal sparger has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 10mm port on 3L metric headplate. L-Shaped Hose Barb Sparger W/SST Filter,3L. Material: 316L with electropolish finish. Overall Length: 11 3/16". Insertion Length: 9 3/4". Supply complete: O-Ring, SST Washer & Nut. SST Filter: 10µm. . MADE IN THE USA
Catalog No. 50-194-6435 Supplier Bellco Glass, Inc. Supplier No. 790900024
Stainless steal sparger has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 10mm port on 3L metric headplate. L-Shaped Hose Barb Sparger W/SST Filter,3L. Material: 316L with electropolish finish. Overall Length: 11 3/16". Insertion Length: 9 3/4". Supply complete: O-Ring, SST Washer & Nut. SST Filter: 10µm. . MADE IN THE USA