Learn More
Bellco Glass, Inc. L-Shape Sparger W/Stainless Steel Frit ,7L
SDP
Catalog No. 501946451
Encompass_Preferred
Stainless steal sparger has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 10mm port on 3L metric headplate. Overall Length: 14.7/16". Insertion Length: 13". Supply complete: O-Ring, SST Washer & Nut.. MADE IN THE USA
Catalog No. 50-194-6451 Supplier Bellco Glass, Inc. Supplier No. 790900045
Stainless steal sparger has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 10mm port on 3L metric headplate. Overall Length: 14.7/16". Insertion Length: 13". Supply complete: O-Ring, SST Washer & Nut.. MADE IN THE USA