Learn More
Alkali Scientific L-Shape Spreaders, 38 x 156mm (10/Sterile Peel, 50 Peels), 500/cs
SDP
Catalog No. 502336214
Encompass_Preferred
These L-shape spreaders are ideal for spreading bacteriological cultures evenly. The shape allows the user to rotate and apply easily throughout an entire round plate or dish. They are made of ABS and have a slight upward curve at the edge to prevent dragging or scraping.
Catalog No. 50-233-6214 Supplier Alkali Scientific Supplier No. TS5421
These L-shape spreaders are ideal for spreading bacteriological cultures evenly. The shape allows the user to rotate and apply easily throughout an entire round plate or dish. They are made of ABS and have a slight upward curve at the edge to prevent dragging or scraping.