Learn More
Grainger L-Shape Wrkbnch ESD Laminate 72 W 30 D
Catalog No. 502768107
Encompass_Preferred
The PRO-LINE L-Shaped Workbench Fixed Height from Grainger is a heavy-duty steel workbench with an L-shaped configuration. It features a 1-1/4" thick maple laminate work surface that measures 60" x 72" and can support up to 2500 lbs. The bench has lockable casters for mobility and adjustable leveling feet for stability. The steel frame and legs have a powder coated finish to resist corrosion. This high-quality workbench provides a large, durable work surface for garage, shop, or industrial use.
Catalog No. 50-276-8107 Supplier Grainger Supplier No. 16Y937
The PRO-LINE L-Shaped Workbench Fixed Height from Grainger is a heavy-duty steel workbench with an L-shaped configuration. It features a 1-1/4" thick maple laminate work surface that measures 60" x 72" and can support up to 2500 lbs. The bench has lockable casters for mobility and adjustable leveling feet for stability. The steel frame and legs have a powder coated finish to resist corrosion. This high-quality workbench provides a large, durable work surface for garage, shop, or industrial use.