Learn More
Bellco Glass, Inc. L-Shaped Cell Spreader, 10/Pack, Sterile
SDP
Catalog No. 502330869
Encompass_Preferred
Flexible for easy access to small volume tubes and dishesColor coded for easy identificationGentle on delicate agar or other surfacesL-Shaped Cell Spreader is ideal for evenly spreading bacterial cultures; angled tip prevents damaging agar surface Pack Description10/WrapCase Quantity500SterileYesNon-pyrogenicYesPrimary ColorGreenPrimary MaterialPolypropyleneRNase/DNase FreeYes
Catalog No. 50-233-0869 Supplier Bellco Glass, Inc. Supplier No. 229617000
Flexible for easy access to small volume tubes and dishesColor coded for easy identificationGentle on delicate agar or other surfacesL-Shaped Cell Spreader is ideal for evenly spreading bacterial cultures; angled tip prevents damaging agar surface Pack Description10/WrapCase Quantity500SterileYesNon-pyrogenicYesPrimary ColorGreenPrimary MaterialPolypropyleneRNase/DNase FreeYes