Learn More
CELLTREAT Scientific Products L-Shaped Cell Spreader, 10/Pack, Sterile
SDP
Catalog No. 50403864
Encompass_Preferred
CELLTREAT Scientific Products L-Shaped Spreader. Flexible for easy access to small volume tubes and dishes. Color coded green for easy identification. Gentle on delicate agar or other surfaces. L-Shaped Cell Spreader is ideal for evenly spreading bacterial cultures; angled tip prevents damaging agar surface. 145mm Length. Made from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Bulk packed sterile, 10/pack, 500/case.
Catalog No. 50-403-864 Supplier CELLTREAT Scientific Products Supplier No. 229617
CELLTREAT Scientific Products L-Shaped Spreader. Flexible for easy access to small volume tubes and dishes. Color coded green for easy identification. Gentle on delicate agar or other surfaces. L-Shaped Cell Spreader is ideal for evenly spreading bacterial cultures; angled tip prevents damaging agar surface. 145mm Length. Made from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Bulk packed sterile, 10/pack, 500/case.