Learn More
Bellco Glass, Inc. L-Shaped Sparger W/Holes,15L For 18mm Port
SDP
Catalog No. 501946479
Encompass_Preferred
Stainless steal L-Shaped Drilled Sparger Assy has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 18mm port on 3L metric headplate. Dimensions: 18 532" L x 5 7/8" bend x 375" OD. Supply with O-Ring, washer, nut, sparger nut. MADE IN THE USA
Catalog No. 50-194-6479 Supplier Bellco Glass, Inc. Supplier No. 790915053
Stainless steal L-Shaped Drilled Sparger Assy has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 18mm port on 3L metric headplate. Dimensions: 18 532" L x 5 7/8" bend x 375" OD. Supply with O-Ring, washer, nut, sparger nut. MADE IN THE USA