Learn More
Bellco Glass, Inc. L-Shaped Sparger W/Holes,3L For 10mm Port
SDP
Catalog No. 501946436
Encompass_Preferred
Stainless steal sparger has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 10mm port on 3L metric headplate. L-Shaped Hose Barb Sparger With Drilled Holes,3L. Material: 316L with electropolish finish. Overall Length: 11 3/16". Insertion Length: 9 3/4". Hole Sizes: 0..06" (7 holes). Supply complete: O-Ring, SST Washer & Nut.. MADE IN THE USA
Catalog No. 50-194-6436 Supplier Bellco Glass, Inc. Supplier No. 790900025
Stainless steal sparger has stainless steal frit with 10 micron porosity for introduction of gentle stream of air bubbles into bioreactor vessel beneath the impeller of mammalian cell culture. Fits into 10mm port on 3L metric headplate. L-Shaped Hose Barb Sparger With Drilled Holes,3L. Material: 316L with electropolish finish. Overall Length: 11 3/16". Insertion Length: 9 3/4". Hole Sizes: 0..06" (7 holes). Supply complete: O-Ring, SST Washer & Nut.. MADE IN THE USA