Learn More
Bellco Glass, Inc. L-Shaped Sparger W/Holes,7L For 10mm Port
SDP
Catalog No. 501946452
Encompass_Preferred
L-Shaped Hose Barb Sparger With Drilled Holes,7L. Fits For 10mm Port on 3L metric headplate. 316L. Overall Length:14 7/16". Insertion Length:13 1/16". Hole Sizes: 0..06" (7 holes). Supply complete: O-Ring, SST Washer & Nut.. MADE IN THE USA
Catalog No. 50-194-6452 Supplier Bellco Glass, Inc. Supplier No. 790900046
L-Shaped Hose Barb Sparger With Drilled Holes,7L. Fits For 10mm Port on 3L metric headplate. 316L. Overall Length:14 7/16". Insertion Length:13 1/16". Hole Sizes: 0..06" (7 holes). Supply complete: O-Ring, SST Washer & Nut.. MADE IN THE USA