Learn More
Copan Diagnostics L-Shaped Spreaders
SDP
Catalog No. 23600899 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Quantity:
1/Peel Pk., 500 Spreaders/Box
10/Peel Pk., 500 Spreaders/Box
5/Peel Pk., 1000 Spreaders/Box
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-600-899 10/Peel Pk., 500 Spreaders/Box
23-600-897 1/Peel Pk., 500 Spreaders/Box
23-600-898 5/Peel Pk., 1000 Spreaders/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-600-899 Supplier Copan Diagnostics Supplier No. 174CS10
Add to Cart
Add to Cart
This item is not returnable. View return policy
L-shaped spreaders
L-Shaped Plastic Spreaders
  • For spreading liquid samples
  • Foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections in the plastic
  • Enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium
  • Available in two sterile peel pack sizes, five or ten spreaders per pack
Specifications
Quantity 10/Peel Pk., 500 Spreaders/Box
Shape L-Shape
Product Type Cell Spreader
For Use With (Equipment) Culture Plates
Sterility Sterile
Material Plastic
For Use With (Application) Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium