Learn More
Grainger L-Shaped Workbench Laminate 72 W 30 D
Catalog No. 502767493
Encompass_Preferred
The Pro-Line L-Shaped Workbench from Grainger is a fixed-height workbench made of steel with a powder-coated finish. It features an L-shaped design with two work surfaces that measure 60"W x 24"D and 48"W x 24"D. The total dimensions are 60"W x 48"D x 34"H. This industrial-grade workbench can support up to 1200 lbs and comes equipped with (2) shelves, (3) drawers, and multiple pegboard storage options for organizing tools and materials. The workbench requires minor assembly and is backed by a 10-year limited warranty from the manufacturer.
Catalog No. 50-276-7493 Supplier Grainger Supplier No. 16Y936
The Pro-Line L-Shaped Workbench from Grainger is a fixed-height workbench made of steel with a powder-coated finish. It features an L-shaped design with two work surfaces that measure 60"W x 24"D and 48"W x 24"D. The total dimensions are 60"W x 48"D x 34"H. This industrial-grade workbench can support up to 1200 lbs and comes equipped with (2) shelves, (3) drawers, and multiple pegboard storage options for organizing tools and materials. The workbench requires minor assembly and is backed by a 10-year limited warranty from the manufacturer.