Learn More
TARGETMOL CHEMICALS INC L-Sodium lactate 200MG
SDP
Catalog No. 502879003
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (L)-Sodium lactate (Sodium L-Lactate) is produced from pyruvate by the enzyme Lactate Dehydrogenase. Lactate production occurs during anaerobic glycolysis or in proliferatively active cells. L-Lactic Acid occurs in small quantities in the blood and muscle fluid of man and animals. The lactic acid concentration increases in muscle and blood after vigorous activity. L-Lactic acid is also present in liver, kidney, thymus gland, human amniotic fluid, and other organs and body fluids. Purity 99.17%
Catalog No. 50-287-9003 Supplier TARGETMOL CHEMICALS INC Supplier No. T4103200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (L)-Sodium lactate (Sodium L-Lactate) is produced from pyruvate by the enzyme Lactate Dehydrogenase. Lactate production occurs during anaerobic glycolysis or in proliferatively active cells. L-Lactic Acid occurs in small quantities in the blood and muscle fluid of man and animals. The lactic acid concentration increases in muscle and blood after vigorous activity. L-Lactic acid is also present in liver, kidney, thymus gland, human amniotic fluid, and other organs and body fluids. Purity 99.17%